Learn More
CD16 Monoclonal Antibody (eBioCB16 (CB16)), Super Bright™ 645, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
182.00 CHF - 396.00 CHF
Spezifikation
Antigen | CD16 |
---|---|
Klon | eBioCB16 (CB16) |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15815643
|
Affymetrix eBioscience
64-0168-42 |
100 Tests |
396.00 CHF
100 Stück |
Verfügbar ab: 10-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15805643
|
Invitrogen
64-0168-41 |
25 Tests |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Description: The eBio1D3 monoclonal antibody reacts with mouse CD19, a 95 kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, MHC class II, and CD19 form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. Applications Reported: This eBio1D3(1D3) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio1D3(1D3) antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No.
CD16 Monoclonal antibody specifically detects CD16 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD16 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
O75015, P08637 | |
FCGR3A, FCGR3B | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
FCGR3A |
eBioCB16 (CB16) | |
Flow Cytometry | |
Super Bright 645 | |
Mouse | |
Human | |
2214, 2215 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |