Learn More
CD16 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16559-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The CD16 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD16. This antibody recognizes human antigen. The CD16 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Spezifikation
| CD16 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 2214 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
| 200 μg/mL | |
| Rabbit | |
| Primary | |
| Human |