missing translation for 'onlineSavingsMsg'
Learn More

CD155 Mouse anti-Human, APC, Clone: 2H7CD155, eBioscience™

Mouse Monoclonal Antibody

Marke:  Invitrogen 17-1550-41

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983204

  • 159.00 CHF / 25 Stück


Verfügbar ab
07-10-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The protein encoded by this gene is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The external domain mediates cell attachment to the extracellular matrix molecule vitronectin, while its intracellular domain interacts with the dynein light chain Tctex-1/DYNLT1. The gene is specific to the primate lineage, and serves as a cellular receptor for poliovirus in the first step of poliovirus replication. Multiple transcript variants encoding different isoforms have been found for this gene.
Spezifikation

Spezifikation

CD155
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P15151
PVR
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
5817
Flow Cytometry
2H7CD155
APC
PVR
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos
Dokumentation