missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse anti-Human, Super Bright 436, Clone: 61D3, eBioscience™

Mouse Monoclonal Antibody

Marke:  Invitrogen 62-0149-42

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983344

  • 306.00 CHF / 100 Stück


Verfügbar ab
09-10-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.
Spezifikation

Spezifikation

CD14
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P08571
CD14
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
61D3
Super Bright 436
CD14
Liquid
CD14
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
929
Videos
Dokumentation