missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Mouse anti-Human, BB700, Clone: MφP9 (also known as MφP-9), BD Horizon™
Mouse Monoclonal Antibody
Marke: BD Biosciences 566466
Additional Details : Gewicht : 0.01000kg
Beschreibung
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD14 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
25 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2b κ |
Flow Cytometry | |
MφP9 (also known as MφP-9) | |
Mouse | |
RUO | |
Human | |
Purified |