missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD134 Mouse anti-Human, BB700, Clone: ACT35 (also known as Ber-ACT35), BD Horizon™
Mouse Monoclonal Antibody
Marke: BD Biosciences 566559
Additional Details : Gewicht : 0.00840kg
Beschreibung
CD134 Monoclonal antibody specifically detects CD134 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD134 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
100 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Flow Cytometry | |
ACT35 (also known as Ber-ACT35) | |
Mouse | |
RUO | |
Human | |
Purified |