missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD133 Mouse anti-Human, APC, Clone: W6B3C1 (also known as W6B3), BD Pharmingen™
Mouse Monoclonal Antibody
Marke: BD Biosciences 566596
Additional Details : Gewicht : 0.00800kg
Beschreibung
CD133 Monoclonal antibody specifically detects CD133 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD133 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
W6B3C1 (also known as W6B3) | |
APC | |
0.1 mg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |