Learn More
CD127 Mouse anti-Human, FITC, Clone: HIL-7R-M21, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 561697
Additional Details : Gewicht : 0.02000kg
Beschreibung
The hIL-7R-M21 monoclonal antibody specifically binds to the 60-90 kDa glycoprotein, CD127. CD127 is also known as the IL-7 receptor alpha (IL-7Rα) subunit. The IL-7 receptor complex is a heterodimer composed of CD127 and the common gamma chain (γc, CD132), shared by other cytokine receptors (IL-2R, IL-4R, IL-9R, IL-15R, and IL-21R). CD127 is expressed on thymocytes, T- and B-cell progenitors, mature T cells, and some lymphoid and myeloid cells. In vitro experiments show the expression of CD127 is down-regulated following T cell activation. Studies indicate that the IL-7 Receptor plays an important role in the proliferation and differentiation of mature T cells. Recently, it has been shown that low surface expression of CD127, in combination with intermediate to high surface expression of CD25, the α chain of the IL-2 receptor complex, can distinguish between human regulatory and conventional CD4+ T cells in human adult and cord blood, lymph nodes and thymus.
Spezifikation
CD127 | |
Monoclonal | |
FITC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human IL-7R Recombinant Protein | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
HIL-7R-M21 | |
IL-7R; IL7R; IL7RA; IL-7Rα; IL-7R-alpha; Interleukin-7 Receptor alpha | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
For Research Use Only.