missing translation for 'onlineSavingsMsg'
Learn More

CD11c Mouse, PE-Cy7, Clone: 3.9, enQuireBio™

Mouse Monoclonal Antibody

184.00 CHF - 346.00 CHF

Spezifikation

Antigen CD11c
Klon 3.9
Konzentration 5μL/Test
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15971718
Unterlagen ansehen
EnquireBio
QAB24-PE7-25Tests
25 Tests
184.00 CHF
25 Tests
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
15961718
Unterlagen ansehen
EnquireBio
QAB24-PE7-100Tests
100 Tests
346.00 CHF
100 Tests
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung

Beschreibung

Spezifikation

Spezifikation

CD11c
5μL/Test
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
3687
IgG1 κ
Protein A/G Purified
3.9
Flow Cytometry
PE-Cyanine7
RUO
XP_011544154
ITGAX
Primary
Store at 2-4°C.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD11c Mouse, PE-Cy7, Clone: 3.9, enQuireBio™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.