missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Hamster anti-Mouse, BV421, Clone: N418, BD Horizon™
Hamster Monoclonal Antibody
Marke: BD Biosciences 565452
Additional Details : Gewicht : 0.01000kg
Beschreibung
CD11c Monoclonal antibody specifically detects CD11c in Mouse samples. It is validated for Flow cytometry.
CD11c Monoclonal antibody specifically detects CD11c in Mouse samples. It is validated for Flow cytometry.Spezifikation
CD11c | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Hamster | |
0.1 mg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
N418 | |
Brilliant Violet 421 | |
Itgax; Integrin alpha-X; Integrin αX; ITAX; CR4; Complement Receptor 4 | |
Mouse Dendritic Cells | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
IgG2 |
For Research Use Only.