missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c (Dendritic Cell Marker) Mouse, Clone: ITGAX/2507, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 3687-MSM6-P0
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD11c (Dendritic Cell Marker) | |
Cell Surface | |
ITGAX/2507 | |
Unconjugated | |
Mouse | |
145kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
3687 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human ITGAX protein (apprx. aa 637-827) | |
THP-1 cells. Tonsil or Lymph Node | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |