missing translation for 'onlineSavingsMsg'
Learn More

CD10 Monoclonal Antibody (eBioCB-CALLA (CB-CALLA)), Super Bright™ 645, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

185.00 CHF - 392.00 CHF

Spezifikation

Antigen CD10
Klon eBioCB-CALLA (CB-CALLA)
Konzentration 5 μL/Test
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15835633
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
64-0106-42
100 Tests
392.00 CHF
100 Stück
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15825633
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
64-0106-41
25 Tests Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
N/A Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Beschreibung

Beschreibung

Description: The M1/70 monoclonal antibody reacts with mouse CD11b, the 165-170 kDa integrin alphaM. CD11b non-covalently associates with CD18 to form alphaM-beta2 integrin (Mac-1) and binds to CD54 (ICAM-1), C3bi, and fibrinogen. Mac-1 is expressed by macrophages, NK cells, granulocytes, activated lymphocytes and mouse B-1 cells in the peritoneal cavity. M1/70 is also cross-reactive to human CD11b, and can be used for the detection of this antigen on human peripheral blood monocytes, granulocytes, and a subset of NK cells. Through interactions with its ligands, CD11b participates in adhesive cell interactions. Applications Reported: This M1/70 antibody has been reported for use in flow cytometric analysis. Applications Tested: This M1/70 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.

CD10 Monoclonal antibody specifically detects CD10 in Human samples. It is validated for Flow Cytometry.
Spezifikation

Spezifikation

CD10
5 μL/Test
Monoclonal
Liquid
RUO
P08473
IgG2b κ
Affinity chromatography
Antibody
eBioCB-CALLA (CB-CALLA)
Flow Cytometry
Super Bright 645
Mouse
Human
4311
Primary
4° C, store in dark, DO NOT FREEZE!
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications