Learn More
PeproTech CCL1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Marke: PeproTech 500-P110BT-25UG
Beschreibung
AA Sequence of recombinant protein: SKSMQVPFSR CCFSFAEQEI PLRAILCYRN TSSICSNEGL IFKLKRGKEA CALDTVGWVQ RHRKMLRHCP SKRK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human I-309 (CCL1). Anti-Human I-309 (CCL1)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human I-309 (CCL1) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human I-309 (CCL1) (500-P110) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human I-309 (CCL1). Western Blot: To detect Human I-309 (CCL1) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human I-309 (CCL1) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P110BT-1MG will be provided as 2 x 500 μg
This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine is secreted by activated T cells and displays chemotactic activity for monocytes but not for neutrophils. It binds to the chemokine receptor CCR8.
Spezifikation
| CCL1 | |
| Polyclonal | |
| Biotin | |
| CCL1 | |
| BF534335; C-C motif chemokine 1; C-C motif chemokine ligand 1; CCL 1; Ccl1; CCR8 ligand; chemokine (C-C motif) ligand 1; I-309; inflammatory cytokine I-309; P500; SCYA1; SISe; SIS-epsilon; small inducible cytokine A1; small inducible cytokine A1 (I-309, homologous to mouse Tca-3); small-inducible cytokine A1; T lymphocyte-secreted protein I-309; TCA3; TCA-3; T-cell activation protein 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6346 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P22362 | |
| CCL1 | |
| E.coli-derived Recombinant Human I-309 (CCL1). | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.