missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC85B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18282-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
DIPA functions as a transcriptional repressor. It may inhibit the activity of CTNNB1 in a TP53-dependent manner and thus regulate cell growth. It may function in adipocyte differentiation, negatively regulating mitotic clonal expansion.Spezifikation
CCDC85B | |
Polyclonal | |
Unconjugated | |
CCDC85B | |
CCDC85B, Delta interacting protein A, DIPA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11007, 240514 | |
Antibody | |
IgG |
Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15834, Q6PDY0 | |
CCDC85B | |
CCDC85B Fusion Protein Ag13129 | |
20 μL | |
Primary | |
Human, Mouse | |
Liquid |