missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin G Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA599402
Additional Details : Gewicht : 0.01000kg
Beschreibung
Antibody detects endogenous levels of total Cathepsin G.
Cathepsin G is a serine protease found in azurophilic granules of neutrophils involved in phagocytic lysis of engulfed forein particles. It is also involved in NK cell cytotoxicity.Spezifikation
Cathepsin G | |
Polyclonal | |
Unconjugated | |
CTSG | |
CATG; cathepsin G; CG; Ctsg; CTSG protein; MGC23078; Vimentin-specific protease; VSP | |
Rabbit | |
Affinity chromatography | |
RUO | |
13035, 1511, 290257 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P08311, P17977, P28293 | |
CTSG | |
A synthesized peptide derived from human CTSG(Accession P08311), corresponding to amino acid residues R96-G146. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |