Learn More
Cathepsin G Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA589049
Additional Details : Gewicht : 0.01000kg
Beschreibung
Immunogen sequence: GAHNIQRREN TQQHITARRA IRHPQYNQRT IQNDIMLLQL SRRVRRNRNV NPVALPRAQE GLRPGTLCTV AGWGRVSMRR GTDTLREVQL RVQRDRQCLR IFGSYDPRRQ ICVGDR; Positive Samples: Mouse kidney, Rat kidney; Cellular Location: Cell surface
Cathepsin G is a serine protease found in azurophilic granules of neutrophils involved in phagocytic lysis of engulfed forein particles. It is also involved in NK cell cytotoxicity.Spezifikation
Cathepsin G | |
Polyclonal | |
Unconjugated | |
CTSG | |
CATG, cathepsin G, CG, Ctsg, CTSG protein, MGC23078, Vimentin-specific protease, VSP | |
Rabbit | |
Affinity Chromatography | |
RUO | |
13035, 1511, 290257 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal, pH 7.3 | |
P08311, P17977, P28293 | |
CTSG | |
Recombinant fusion protein containing a sequence corresponding to amino acids 75-190 of human Cathepsin G (NP_0019021) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |