missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catalase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21260-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kD in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara′s disease.Spezifikation
Catalase | |
Polyclonal | |
Unconjugated | |
CAT | |
CAT, catalase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12359, 24248, 847 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04040, P04762, P24270 | |
CAT | |
Catalase Fusion Protein Ag15383 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |