Learn More
Caspase 1 (Cleaved Asp210) Polyclonal Antibody, Invitrogen™
Artikelnummer. 15515031
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15515031 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15515031 Lieferant Invitrogen Lieferanten-Nr. PA538100
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Rabbit Polyclonal Antibody
Caspases are a family of cysteine proteases that are key mediators of programmed cell death or apoptosis (1). The precursor form of all caspases is composed of a prodomain, and large and small catalytic subunits. The active forms of caspases are generated by several stimuli including ligand-receptor interactions, growth factor deprivation and inhibitors of cellular functions. All known caspases require cleavage adjacent to aspartates to liberate one large and one small subunit, which associate into a tetramer to form the active enzyme. Caspase-1/ICE (IL-1b converting enzyme) is similar to the cell death gene CED-3 of Caenorhabditilis elegans and regulates multiple proinflammatory cytokines, including interleukin-1b and interferon-gamma-inducing factor.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Caspase 1 (Cleaved Asp210)
Anwendungen Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gen CASP1
Gen-Zugriffsnummer P29466
Gen-Alias CASP-1, ICE, IL-1 beta converting enzyme, IL-1BC, IL1BCE, caspase-1, interleukin-1 beta convertase, interleukin-1 beta converting enzyme, p45
Gensymbole CASP1
Wirtsspezies Rabbit
Immunogen A synthetic peptide derived from human Caspase 1 (MTTELEAFC)
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 834
Zielspezies Human
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG