Learn More
Cas9 Monoclonal Antibody (10C11-A12), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA1202
457.60 CHF gültig bis 2024-06-28
Benutzen Sie den Promotions-Code "21649" um Ihren Promotions-Preis zu erhalten.
Additional Details : Gewicht : 0.01000kg
Beschreibung
Cas9 (10C11-A12) Mouse mAb recognizes transfected levels of total Cas9 protein. Stable Cas9 U2OS cell line used in validation experiments expresses Cas9 at lower levels. Assay results may vary depending on expression level of Cas9 in cell lines tested. This product was tested in Streptococcus pyogenes bacteria.
Cas9 is an RNA-guided DNA endonuclease enzyme associated with the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) adaptive immunity system in bacteria and genetic engineering. The system is unique and flexible due to its dependence on RNA as the element that targets the nuclease to a desired DNA sequence. It can be used to induce indel mutations, specific sequence replacements/insertions, and large deletions or genomic rearrangements at any desired location in the genome. In addition, Cas9 can also be used to mediate upregulation of specific endogenous genes or to alter histone modifications or DNA methylation. Cas9 may be derived from Streptococcus pyogenes (SpCas9) or Staphylococcus aureus (SaCas9). SaCas9 has a number of properties that make it advantageous for genome editing, including its small size, high efficiency, nickase activity, and apparent specificity. SaCas9 principally recognizes an NNGRRT protospacer adjacent motif (PAM) and cleaves target DNA at rates comparable to SpCas9 with an ideal spacer lengths of 24 to 20 nucleotides. In comparison to SpCas9, SaCas9 is smaller (1368 AA versus 1053 AA) and generates indels at a comparable rate when directed against target DNA with the mutually permissive NGGRRT PAM. This makes it the preferred enzyme for all-in-one delivery of Cas9 and multiple gRNA expression cassettes with Adeno-associated vectors.Spezifikation
Cas9 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
Q99ZW2 | |
cas9 | |
Cas9 nuclease | |
100 μg | |
Primary | |
Bacteria | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunocytochemistry, Western Blot | |
10C11-A12 | |
Unconjugated | |
cas9 | |
cas9; CASP 9; Caspase9; Caspase-9 precursor; CASPASE-9c; CRISPR9; CRISPR-Cas9; CTD; OTTHUMP00000002324; RNCASP9; RP11-265F14.3; s aureus | |
Mouse | |
Protein G | |
RUO | |
901176 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |