missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carvedilol

Catalog Number p-7214416
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
72956-09-3
C24H26N2O4
406.48
MFCD00864692,MFCD00869663
OGHNVEJMJSYVRP-UHFFFAOYNA-N
1-(9H-Carbazol-4-Yloxy)-3-{[2-(2-Methoxyphenoxy)ethyl]amino}Propan-2-ol
COC1=CC=CC=C1OCCNCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
72956-09-3
C24H26N2O4
406.48
MFCD00864692,MFCD00869663
OGHNVEJMJSYVRP-UHFFFAOYNA-N
1-(9H-Carbazol-4-Yloxy)-3-{[2-(2-Methoxyphenoxy)ethyl]amino}Propan-2-ol
COC1=CC=CC=C1OCCNCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Carvedilol
Schmelzpunkt 114.0°C to 115.0°C
Menge 1 g
Löslichkeitsinformationen Solubility aq. soln.: insoluble. Other solubilities: very soluble in dimethyl sulfoxide, soluble in methanol & methylene chloride, slightly soluble in ether, ethanol & isopropanol
Formelmasse 406.47

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'