Learn More
Carpenter 20 Gaze, 12 Mesh gewebt aus Draht mit Durchmesser von 0.58 mm (0.023 Zoll), Thermo Scientific™
Marke: Thermo Scientific Alfa Aesar 046526.HB
Additional Details : Gewicht : 0.01000kg
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
Carpenter 20 gauze | |
300 x 300 mm | |
Odorless |
12 mesh woven from 0.58mm (0.023 in.) dia. wire | |
12 Mesh | |
Drahtgewebe |