Learn More
Cardiac Troponin T Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 15513-1-AP-20UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16814724
  • 136.48 CHF / 20 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Cardiac Troponin T is the tropomyosin-binding subunit of the troponin complex, which is located on the thin filament of striated muscles and regulates muscle contraction in response to alterations in intracellular calcium ion concentration. Cardiac Troponin T is important in proper muscle function and myofibril formation. In the human heart, four cTnT isoforms are expressed in a developmentally regulated manner. Cardiac Troponin T expression in the human heart is affected by heart failure. Mutations within the Cardiac Troponin T protein have been associated with familial hypertrophic cardiomyopathy as well as with dilated cardiomyopathy. Transcripts for the Cardiac Troponin T gene undergo alternative splicing that results in many tissue-specific isoforms.
Spezifikation
Spezifikation
Cardiac Troponin T
Polyclonal
Unconjugated
TNNT2
Cardiac Troponin T, CMH2, cTnT, TNNT2, TnTC
Rabbit
Antigen Affinity Chromatography
RUO
21956, 24837, 7139
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P45379, P50752, P50753
TNNT2
Cardiac Troponin T Fusion Protein Ag7716
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen