Learn More
Paragon Scientific™ Cannon-Fenske Routine Viscometer for Transparent Liquids (UKAS ISO 17025 certified)
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
Marke:  Paragon Scientific™ CFR03UKC
Additional Details : Gewicht : 0.40000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15909999
  • 197.00 CHF / 1 Stück
Lager auf Anfrage
056 618 41 11
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
  • Supplied with full UKAS ISO 17025 calibration
Spezifikation
Spezifikation
D 445, D 446
Yes
UKAS ISO 17025 calibration
1.6 to 8 cSt
0.008
ASTM D 445, D 446 and ISO 3104, 3105
75
Videos
Dokumentation
8 week lead-time