missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Canine TNF-alpha Recombinant Protein

Artikelnummer. 11898761
Click to view available options
Menge:
20 μg
Packungsgröße:
20 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11898761

Marke: Invitrogen™ RCTNFAI

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

For dilution, use a buffer containing carrier protein, e.g. PBS with 1% BSA or a neutral media containing 10% FCS and freeze in aliquots at -70°C. Activity: The biological activity of recombinant canine TNFa was measured by its cytolytic effect on mouse WEHI-13VAR cells in the presence of Actinomycin D. The ED50 (or 1 unit of activity) for this effect in our laboratories is typically 10 pg/mL. Refs: Kitani, H. et al. (2002) Infect. Immun. 70:2210. Khabar, K.S. et al. (1995) Immunol. Lett. 46:107. Expression: The mature, soluble form of canine TNF-alpha was expressed in Pichia pastoris, X-33. GenBank™ Accession # P51742, amino acids 77 - 233. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: VKSSSRTPSD. Reconstitute with 0.5 mL of distilled water.

TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P51742
Zur Verwendung mit (Anwendung) ELISA, Immunoprecipitation, Western Blot
Zusammensetzung PBS with no preservative
Gen-ID (Entrez) 403922
Molekulargewicht 17.3 kDa
Name Canine TNF-alpha
Menge 20 μg
Quelle Yeast
Kennzeichnung RUO
Gen-Alias APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF a; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFa; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1 F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor
Gebräuchliche Bezeichnung TNF alpha
Gensymbol TNF
Biologische Aktivität ED50 = 10 pg/mL; determined by the dose-dependent cytolytic effect on mouse WEHI-13 VAR cells in the presence of Actinomycin D.
Konjugat Unconjugated
Spezies Yeast
Rekombinant Recombinant
Sequenz Canine TNF-alpha, amino acids 77-233 (Accession # P51742)
Inhalt und Lagerung 4°C
Expressionssystem P. pastoris
Form Lyophilized
Protein-Subtyp Recombinant
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt