Learn More
R&D Systems™ Canine CCR5 (NP_001012342) VersaClone cDNA
Beschreibung
The Canine CCR5 (NP_001012342) VersaClone cDNA clone contains the complete ORF for CCR5, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CCR5 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Bestellinformation
Ships at ambient temperature.
Spezifikation
Spezifikation
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Produktlinie | R&D Systems |
| Menge | 10 μg |
For Reasearch Use Only; Not for Use in Humans