missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALR3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17064-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Calreticulins, such as Calreticulin 3, are Ca(2+)-binding chaperones localized mainly in the endoplasmic/sarcoplasmic reticulum.Spezifikation
| CALR3 | |
| Polyclonal | |
| Unconjugated | |
| CALR3 | |
| CALR3, Calreticulin 2, calreticulin 3, CRT2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 125972, 364529, 73316 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96L12, Q9D9Q6 | |
| Calr3 | |
| CALR3 Fusion Protein Ag10847 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction