missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calnexin (Endoplasmic Reticulum Marker) Mouse, Clone: CANX/1543, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 821-MSM3-P1
Additional Details : Gewicht : 0.01000kg
Spezifikation
Calnexin (Endoplasmic Reticulum Marker) | |
Cytoplasmic | |
CANX/1543 | |
Unconjugated | |
Mouse | |
90kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
821 | |
IgG1 κ |
ELISA, Western Blot, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant N-terminal fragment of human Calnexin protein (apprx. aa 1-300) | |
HeLa, MCF-7 or U2OS cells. Kidney or small intestine. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |