Learn More
Unity Lab Services™ Calibration Certificate Cofrac Temperature Two Fixed Points, 0°C, 120°C
Artikelnummer. 12655670
Ansicht ändern
Click to view available options
Produkttyp:
Kalibrierungszertifikat
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Produkttyp Packungsgröße
12655670 Kalibrierungszertifikat 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 12655670 Lieferant Unity Lab Services™ Lieferanten-Nr. UMVP935A
 Eingeschränkter Artikel
Verfügbar ab: 20-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
ULS offers a full range of support services and programs for all instruments and supplies, regardless of manufacturer. Maximize your technology investments by anticipating and avoiding problems before they occur.
  • Calibration by comparison with a reference thermometer in baths
  • Type of device: temperature measuring thermometric chain or expansion thermometer, at fixed values of 0°C, 120°C
  • Our best uncertainties for a thermometer with a resolution of 0.01°C (k = 2) ±0.02°C at 0°C; ±0.08°C to 120°C
  • Our best uncertainties for a thermometer with a resolution of 0.1°C (k = 2) ±0.1°C to 0°C; ±0.2°C to 120°C
  • COFRAC Accreditation Number 2-5984
  • Calibration, scope available on www.cofrac.fr
  • Average time: 1 week
TRUSTED_SUSTAINABILITY
Spezifikation
Zur Verwendung mit (Geräte) Thermometers and Temperature Recorders
Zertifizierung/Konformität Cofrac
Dauer 1 Day
Produkttyp Kalibrierungszertifikat