missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNG5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18498-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
L-type calcium channels are composed of five subunits. The protein Calcium Channel 5 represents one of these subunits, gamma, and is one of several gamma subunit proteins. It is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state.Spezifikation
CACNG5 | |
Polyclonal | |
Unconjugated | |
CACNG5 | |
CACNG5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27091 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UF02 | |
CACNG5 | |
CACNG5 Fusion Protein Ag13339 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |