missing translation for 'onlineSavingsMsg'
Learn More

CACNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

136.48 CHF - 385.92 CHF

Spezifikation

Antigen CACNB1
Konzentration 0.27 mg/mL
Anwendungen Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16841054
Unterlagen ansehen
Proteintech
13039-1-AP-20UL
20 μL
136.48 CHF
20 Mikroliter
Nur noch null auf Lager
Lager vorrätig
Zum Warenkorb hinzufügen
 
16831054
Unterlagen ansehen
Proteintech
13039-1-AP-150UL
150 μL
385.92 CHF
150 Mikroliter
Nur noch null auf Lager
Lager vorrätig
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

The expression of multiple classes of voltage dependent calcium channels (VDCC′s) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCC′s exist before birth which is consistent with a role in migration, most N-Type VDCC′s subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
Spezifikation

Spezifikation

CACNB1
Western Blot
Unconjugated
Rabbit
Rat, Human, Mouse
P54283, Q02641, Q8R3Z5
12295, 50688, 782
CACNB1 Fusion Protein Ag3693
Primary
-20°C
CACNB1
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CAB1, CACNB1, CACNLB1, CCHLB1
Cacnb1
IgG
Antigen Affinity Chromatography
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
CACNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.