Learn More
C8orf33 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555292
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RNRASVANGG EKASEKLAPE EVPLSAEAQA QQLAQELAWC VEQLELGLKR QKPTPKQKEQ AIGAIRTLRS K Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 73%.
The function of this protein remains unknown.Spezifikation
C8orf33 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H7E9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
65265 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
C8orf33 | |
C8ORF33 | |
Recombit protein corresponding to Human C8orf33 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only