missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ C6orf166 Recombinant Protein
Marke: Abnova™ H00055122-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH00764 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
48.07 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AKIRIN2 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
55122 | |
C6orf166 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
C6orf166/FBI1/FLJ10342/dJ486L4.2 | |
AKIRIN2 | |
Wheat Germ (in vitro) | |
GST |