missing translation for 'onlineSavingsMsg'
Learn More

C5 Mouse anti-Human, Clone: 2G8C9, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66634-1-IG-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838695

  • 351.68 CHF / 150 Mikroliter

Verfügbar ab: 28-06-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

C5 is part of the membrane attack complex (MAC) of the complement system. C5 is split into C5a og C5b by the C5 convertases from the classical and the alternative complement pathway and the C5b fragment initiates the assembly of the terminal MAC (C5b-C9). The molecular mass of C5 is 190 kDa.
Spezifikation

Spezifikation

C5
Monoclonal
0.93 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P01031
C5
C5 Fusion Protein Ag17882
150 μL
Primary
Human
Liquid
Western Blot
2G8C9
Unconjugated
C5
C5, Complement C5, complement component 5, CPAMD4
Mouse
Protein A
RUO
727
Antibody
IgG2a
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote