Learn More
C3orf26 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560138
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DEWWENQPTG AGSSPEASDG EGEGDTEVMQ QETVPVPVPS EKTKQPKECF LIQPKERKEN TTKTRKRRKK KITDVLAKSE PKP Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 54%.
CMSS1 (Cms1 Ribosomal Small Subunit Homolog) is a Protein Coding gene. Diseases associated with CMSS1 includeRefractive Error. Gene Ontology (GO) annotations related to this gene includenucleic acid binding.Spezifikation
C3orf26 | |
Polyclonal | |
Unconjugated | |
CMSS1 | |
1110001A06Rik; 2610528E23Rik; 4930572F24Rik; AV099812; AV122629; C3orf26; cms small ribosomal subunit 1; cms1 ribosomal small subunit homolog; cms1 ribosomal small subunit homolog (yeast); Cmss1; MGC4308; Protein CMSS1; RGD1309437 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84319 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BQ75 | |
CMSS1 | |
Recombinant protein corresponding to Human C3orf26 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only