missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf92 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21031-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
LRRC71 is a protein coding gene.Spezifikation
C1orf92 | |
Polyclonal | |
Unconjugated | |
LRRC71 | |
C1orf92, LRRC71, RP11 356J7.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
149499 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N4P6 | |
LRRC71 | |
C1orf92 Fusion Protein Ag15344 | |
150 μL | |
Primary | |
Human | |
Liquid |