missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf57 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21463-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Belonging to the THEP1 NTPase family, C1orf57 (also known as nucleoside triphosphate phosphohydrolase), and its mouse homolog, 2310079N02Rik, are 190 amino acid proteins that has nucleotide phosphatase activity towards ATP, GTP, TTP, CTP and UTP. Acting as a monomer, it also hydrolyzes nucleoside diphosphates with lower efficiency. The gene encoding C1orf57 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1.Spezifikation
C1orf57 | |
Polyclonal | |
Unconjugated | |
NTPCR | |
C1orf57, NTPase, RP4 678E16.2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361443, 66566, 84284 | |
Antibody | |
IgG |
Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BSD7, Q9CQA9 | |
Ntpcr | |
C1orf57 Fusion Protein Ag15009 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |