missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C13orf18 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21183-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Spezifikation
C13orf18 | |
Polyclonal | |
Unconjugated | |
RUBCNL | |
C13orf18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80183 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.48 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H714 | |
RUBCNL | |
C13orf18 Fusion Protein Ag15474 | |
150 μL | |
Primary | |
Human | |
Liquid |