missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C11orf49 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20195-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Spezifikation
C11orf49 | |
Polyclonal | |
Unconjugated | |
C11orf49 | |
C11orf49, UPF0705 protein C11orf49 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
295930, 79096 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H6J7 | |
C11orf49, RGD1309540 | |
C11orf49 Fusion Protein Ag14123 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |