Learn More
IKA™ C1 Calorimeter Package with C1.12 Combustion Chamber
The C1 Calorimeter Package with C1.20 Combustion Chamber combines a compact design with a high degree of automation, featuring a light, halogen-resistant combustion chamber.
Marke:  IKA™ 0010005637
Additional Details : CAS : 65-85-0 Gewicht : 2.30000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 17277479
  • 21165.00 CHF / 1 Stück
Verfügbar ab: 01-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Features a high degree of automation
  • Compact design offers a small footprint
  • Operates in accordance with DIN 51900 and ISO 1928
  • PCI / PCS measurement analysis follows classic isoperibolic methodology
  • Easily retractable combustion chamber with active catalytic coating and halogen-resistance is specially designed for the combustion of sample materials containing halogens and sulfur (>3%)
  • Compatible with IKA™ C6040 Calwin calorimeter software (sold separately), which allows additional extensions and adaptations, such as the management of measurement data and LIMS
  • Includes wear parts and consumables for the first 500 tests including 25 calibrations
  • C1.10 combustion chamber is fitted with a large C5010.5 crucible support and a large C6 quartz crucible
Spezifikation
Spezifikation
Calorimeter
50/60 Hz
Up to 40,000 J
100/240 V
DIN 51900, ISO 1928
C1 Calorimeter, C1.12 Halogen-resistant Combustion Chamber (Requires suitable water cooling supply to operate)
20°C to 30°C