missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Butralin Monoclonal Antibody (3B4)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA549730
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Antibody tested in indirect ELISA.
Butralin is a selective herbicide that is widely used to control broadleaf weeds in various crops, including soybeans, peanuts, cotton, and fruit trees. Its active ingredient, butralin, belongs to the dinitroaniline chemical family, which inhibits the microtubule assembly in plant cells, leading to the disruption of cell division and growth. Butralin can be applied to the soil before or after planting, and it has residual activity that can provide long-lasting weed control.
Spezifikation
| Butralin | |
| Monoclonal | |
| 3 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Conjugate of BTL hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 3B4 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur