missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 8.00, NIST, Chem-Lab
Click to view available options
Menge:
10 L
Verpackung:
PE
Beschreibung
pH = 8.18 at 0°C
pH = 8.09 at 10°C
pH = 8.00 at 20°C
pH = 7.97 at 25°C
pH = 7.94 at 30°C
pH = 7.90 at 40°C
pH = 7.86 at 50°C
pH = 7.82 at 60°C
pH = 7.80 at 70°C
pH = 7.77 at 80°C
pH = 7.75 at 90°C
pH = 8.09 at 10°C
pH = 8.00 at 20°C
pH = 7.97 at 25°C
pH = 7.94 at 30°C
pH = 7.90 at 40°C
pH = 7.86 at 50°C
pH = 7.82 at 60°C
pH = 7.80 at 70°C
pH = 7.77 at 80°C
pH = 7.75 at 90°C
Spezifikation
Dichte | 1.01g/mL |
pH | 8.00 |
Menge | 10 L |
Zusammensetzung | H3BO3, KCl, NaOH |
Verpackung | PE |
Rückführbarkeit auf NIST | NIST traceable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'