missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffer Set C2/C3 Bottle of 100 ml Buffer C2 and 25 ml Buffer C3

Artikelnummer. 12870911
Click to view available options
Menge:
125 mL
Packungsgröße:
1 Set
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 12870911

Marke: Macherey-Nagel™ 740935

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Buffer C2 and Buffer C3, for Buffer 125 mL Buffer C4.

Specifications

Menge 125 mL
compliance-icons
Signal Word
  • Achtung
Hazard Category
  • Akute Toxizität Kategorie 4
  • Schwere Augenschädigung/Augenreizung Kategorie 2
  • Hautreizende/-ätzende Wirkung Kategorie 2
Hazard Statement
  • H302-Gesundheitsschädlich bei Verschlucken.
  • H315-Verursacht Hautreizungen.
  • H319-Verursacht schwere Augenreizung.
Supplemental information
  • MIXTURE LIST-Enthält: guanidine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.