missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BST-2 Monoclonal Antibody (K29052_18D3), Invitrogen™
Mouse Monoclonal Antibody
420.00 CHF - 632.00 CHF
Spezifikation
| Antigen | BST-2 |
|---|---|
| Klon | K29052_18D3 |
| Konzentration | 1 mg/mL |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
30204056
|
Invitrogen™
MA556177 |
200 μg |
632.00 CHF
200 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30203358
|
Invitrogen™
MA555797 |
50 μg |
420.00 CHF
50 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Description
Sequence of this protein is as follows: QQELTEAQKG FQDVEAQAAT CNHTVMALMA SLDAEKAQGQ KKVEELEGEI TTLNHKLQDA SAEVERLRRE NQVLSVRIAD KKYYPSSQDS
BST-2 Monoclonal antibody specifically detects BST-2 in Human samples. It is validated for Western BlotSpecifications
| BST-2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide | |
| 2310015I10Rik; bone marrow stromal antigen 2; bone marrow stromal cell antigen 2; Bst2; BST-2; C87040; CD317; Damp1; DAMP-1; DAMP-1 protein homolog; GREG; HM1.24 antigen; NPC-A-7; PDCA-1; protein DAMP-1; TETHERIN | |
| Recombinant human BST2 (immunogen range is 71-160). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Bst2 |
| K29052_18D3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q10589 | |
| 684 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title