Learn More
Thermo Scientific Chemicals Bradykinin-Acetat-Salz
Beschreibung
Bradykinin Acetate elicits vasoconstriction, a systemic blood pressure reflex, a stimulation of respiration, and contraction of the nictitating membrane. Bradykinin plays an important role in the regulation of fluid and electrolyte balance, smooth muscle contraction, vasodilation and capillary permeability.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Reinigungsverfahren | HPLC |
| Inhalt und Lagerung | -30°bis -10 °C |
| Format | Pulver |
| Molekulargewicht | 1120.21 |
| Produkttyp | Bradykinin acetate salt |
| Menge | 100 mg |
| Kennzeichnung | RUO - research use only |
| Sequenz | Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg-OH |
Nur für Forschungszwecke