Learn More
Invitrogen™ Bolt™ 4 bis 12 %, Bis-Tris, 1,0 mm, Mini-Protein-Gele
Beschreibung
Benötigt
Mini Gel Tank (Herst.- Nr. A25977)
Invitrogen Bolt Bis-Tris Plus Gele sind Polyacrylamid-Fertiggele zur optimalen Trennung von Proteinen verschiedenster Größen unter denaturierenden Bedingungen. Ähnlich wie NuPAGE Bis-Tris-Gele erzielen Bolt Bis-Tris Plus-Gele eine gleichbleibende Gelleistung und sorgen für eine pH-neutrale Umgebung, die Proteinveränderungen bei höherer Beladungskapazität minimiert. Bolt Gele eignen sich hervorragend für Western-Blot-Transfers und -Analysen und für andere Verfahren, bei denen die Unversehrtheit der Proteine entscheidend ist.
Use Bolt Bis-Tris Plus gels to obtain optimal results for your day-to-day protein separation needs. Bolt Bis-Tris Plus gels come in four gel types and multiple well formats. Each box contains 10 gels. A Mini Gel Tank (A25977) is required for use of all Bolt gels.
Features include:
- Optimized run conditions—separate your proteins using constant voltage in approximately 20 minutes
- Long shelf life—store gels for at least 16 months at 4–25°C
- Better protein integrity—neutral pH sample preparation process preserves your proteins
- Wedge-shaped wells—for easier sample loading and up to two times higher sample volume
Choose the right Bolt Bis-Tris Plus gels for your protein separation
Obtain optimal separation of your proteins by choosing the right combination of gel and running buffer. Bolt Bis-Tris Plus gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and 4–12% gradient. The gels can be run using Bolt MES SDS (B000202) or Bolt MOPS SDS (B000102) running buffer to obtain different separation ranges. Bolt Bis-Tris Plus gels consist of a neutral pH-buffered (pH 6.4) polyacrylamide gel and have an operating pH of about 7.0. Bolt Bis-Tris Plus gels also come in four different well formats: 10, 12, 15, and 17 wells.
Convenient to use
Bolt Bis-Tris Plus gels can be run using constant voltage in approximately 20 minutes with the Bolt MES running buffer. Bolt Bis-Tris Plus gels offer sharper bands, high resolution, longer shelf life, and higher accuracy of results than Tris-glycine–based gels. In addition, Bolt Bis-Tris Plus gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0-mm thick gels. The wedge-shaped wells also have larger openings, making sample loading easier. And Bolt Bis-Tris Plus gels have a shelf life of at least 16 months at 4°C to 25°C.
For transferring your proteins to a membrane, we recommend using for Bolt Transfer buffer (BT0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).
Spezifikation
Spezifikation
| Farbe | Clear |
| Gelanteil (%) | 4 to 12% |
| Gelgröße | Mini |
| Gel Thickness | 1.0 mm |
| Geltyp | Bis-Tris Plus |
| Probenladevolumen | Up to 60 μL |
| Trennung von | Protein |
| Trennbereich | 3.5 to 260 kDa |
| Trennverfahren | Denaturing |
| Wells | 15-well |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.