Learn More
Bisphenol A (3-Chloro-2-hydroxypropyl) Glycidyl Ether, TRC
Artikelnummer. 30137284
Change view
Click to view available options
Menge:
10 mg
Packungsgröße:
10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30137284 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 30137284 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCB51967010MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 13836-48-1
Summenformel C21 H25 Cl O4
Molekulargewicht (g/mol) 376.874
Synonym BADGE.HCl, 2-Propanol, 1-chloro-3-[4-[1-methyl-1-[4-(2-oxiranylmethoxy)phenyl]ethyl]phenoxy]-, 2-Propanol, 1-chloro-3-[4-[1-methyl-1-[4-(oxiranylmethoxy)phenyl]ethyl]phenoxy]- (9CI), 2-Propanol, 1-chloro-3-[[α-[p-(2,3-epoxypropoxy)phenyl]-p-cumenyl]oxy]- (8CI), Bisphenol A (3-chloro-2-hydroxypropyl)glycidyl ether, 1-Chloro-3-[4-[1-methyl-1-[4-(2-oxiranylmethoxy)phenyl]ethyl]phenoxy]-2-propanol, BADGE.HCl
IUPAC-Name 1-chloro-3-[4-[2-[4-(oxiran-2-ylmethoxy)phenyl]propan-2-yl]phenoxy]propan-2-ol
SMILES CC(C)(c1ccc(OCC(O)CCl)cc1)c2ccc(OCC3CO3)cc2
Spezifikation
Chemischer Name oder Material Bisphenol A (3-Chloro-2-hydroxypropyl) Glycidyl Ether
Hinweise zur Reinheitsqualität HPLC
Menge 10 mg
InChI-Formel InChI=1S/C21H25ClO4/c1-21(2,15-3-7-18(8-4-15)24-12-17(23)11-22)16-5-9-19(10-6-16)25-13-20-14-26-20/h3-10,17,20,23H,11-14H2,1-2H3
Formelmasse 376.144
Reinheit (%) >95%