Learn More
ACROBiosystems Biotinylated Zika virus NS1 Protein, Avitag™,His Tag
Biotinylated Protein
441.35 CHF - 1941.94 CHF
Spezifikation
Zugriffsnummer | ALU33341.1 |
---|---|
Molekulargewicht | 43.3 kDa |
Quelle | Biotinylated Zika virus NS1, Avitag,His Tag (NS1-Z82E9) is expressed from human 293 cells (HEK293). It contains AA Val 796 - Ser 1148 (Accession # ALU33341.1). |
Immunogen | Val 796 - Ser 1148 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15817377
|
ACROBiosystems
NS1-Z82E9-25UG |
25 μg |
441.35 CHF
25 Mikrogramm |
Verfügbar ab: 02-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15807377
|
ACROBiosystems
NS1-Z82E9-200UG |
200 μg |
1941.94 CHF
200 Mikrogramm |
Verfügbar ab: 02-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Zika virus (ZIKV) is a member of the virus family Flaviviridae and the genus Flavivirus, transmitted by daytime-active Aedes mosquitoes, such as A. aegypti and A. albopictus. Its name comes from the Zika Forest of Uganda, where the virus was first isolated in 1947. Zika virus is related to dengue, yellow fever, Japanese encephalitis, and West Nile viruses. The infection, known as Zika fever, often causes no or only mild symptoms, similar to a mild form of dengue fever. It is treated by rest. Since the 1950s, it has been known to occur within a narrow equatorial belt from Africa to Asia. As of 2016, the illness cannot be prevented by drugs or vaccines. As of February 2016, there is evidence that Zika fever in pregnant women is associated with abnormal brain development in their fetuses through mother-to-child transmission of the virus, which may result in miscarriage or microcephaly.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 43.3 kDa. The protein migrates as 46-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
ALU33341.1 | |
Biotinylated Zika virus NS1, Avitag,His Tag (NS1-Z82E9) is expressed from human 293 cells (HEK293). It contains AA Val 796 - Ser 1148 (Accession # ALU33341.1). | |
-20°C | |
NS1 | |
Zika virus | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
43.3 kDa | |
Val 796 - Ser 1148 | |
NS1 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |