missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Zika virus NS1 Protein, Avitag™,His Tag

Biotinylated Protein

Marke:  ACROBiosystems NS1-Z82E9-25UG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15817377

  • 403.01 CHF / 25 Mikrogramm


Verfügbar ab
04-06-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Zika virus (ZIKV) is a member of the virus family Flaviviridae and the genus Flavivirus, transmitted by daytime-active Aedes mosquitoes, such as A. aegypti and A. albopictus. Its name comes from the Zika Forest of Uganda, where the virus was first isolated in 1947. Zika virus is related to dengue, yellow fever, Japanese encephalitis, and West Nile viruses. The infection, known as Zika fever, often causes no or only mild symptoms, similar to a mild form of dengue fever. It is treated by rest. Since the 1950s, it has been known to occur within a narrow equatorial belt from Africa to Asia. As of 2016, the illness cannot be prevented by drugs or vaccines. As of February 2016, there is evidence that Zika fever in pregnant women is associated with abnormal brain development in their fetuses through mother-to-child transmission of the virus, which may result in miscarriage or microcephaly.

This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 43.3 kDa. The protein migrates as 46-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

ALU33341.1
HEK293
Zika virus
43.3 kDa
25 µg
Val 796 - Ser 1148
NS1
Precision Avi
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
Powder
>90% as determined by SDS-PAGE
Biotinylated Zika virus NS1, Avitag,His Tag (NS1-Z82E9) is expressed from human 293 cells (HEK293). It contains AA Val 796 - Ser 1148 (Accession # ALU33341.1).
-20°C
NS1
His Tag and Avi Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation