Learn More
ACROBiosystems Biotinylated Mouse TNF-alpha Protein, His,Avitag™ (active trimer)
MALS verified
441.35 CHF - 1941.94 CHF
Spezifikation
Molekulargewicht | 20.2 kDa |
---|---|
Quelle | MABSol™ Biotinylated Mouse TNF-alpha, His,Avitag (active trimer) (MALS verified) (TNA-M82E9) is expressed from human HEK293 cells. It contains AA Leu 80 - Leu 235 (Accession # P06804). |
Immunogen | Leu 80 - Leu 235 |
Lagerungsbedingungen | -20°C |
Gen-Alias | DIF,TNF-alpha,TNFA,TNFSF2,cachexin,cachectin,TNFα |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15807847
|
ACROBiosystems
TNA-M82E9-200UG |
200 μg |
1941.94 CHF
200 Mikrogramm |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15817847
|
ACROBiosystems
TNA-M82E9-25UG |
25 μg |
441.35 CHF
25 Mikrogramm |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Tumor necrosis factor alpha (TNFα) is a cytokine produced primarily by monocytes and macrophages. It is found in synovial cells and macrophages in the tissues.The primary role of TNFα is in the regulation of immune cells. TNFα is able to induce apoptotic cell death, to induce inflammation, and to inhibit tumorigenesis and viral replication. Dysregulation of TNFα production has been implicated in a variety of human diseases, including major depression, Alzheimer's disease and cancer. Recombinant TNFα is used as an immunostimulant under the INN tasonermin. TNFα can be produced ectopically in the setting of malignancy and parallels parathyroid hormone both in causing secondary hypercalcemia and in the cancers with which excessive production is associated.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 20.2 kDa. The protein migrates as 20-21 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
20.2 kDa | |
Leu 80 - Leu 235 | |
DIF,TNF-alpha,TNFA,TNFSF2,cachexin,cachectin,TNFα | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
MABSol™ Biotinylated Mouse TNF-alpha, His,Avitag (active trimer) (MALS verified) (TNA-M82E9) is expressed from human HEK293 cells. It contains AA Leu 80 - Leu 235 (Accession # P06804). | |
-20°C | |
TNF-alpha | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |