missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Mouse TNF-alpha Protein, His,Avitag™ (active trimer)

MALS verified

Marke:  ACROBiosystems TNA-M82E9-25UG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15817847

  • 441.35 CHF / 25 Mikrogramm
Verfügbar ab: 17-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Tumor necrosis factor alpha (TNFα) is a cytokine produced primarily by monocytes and macrophages. It is found in synovial cells and macrophages in the tissues.The primary role of TNFα is in the regulation of immune cells. TNFα is able to induce apoptotic cell death, to induce inflammation, and to inhibit tumorigenesis and viral replication. Dysregulation of TNFα production has been implicated in a variety of human diseases, including major depression, Alzheimer's disease and cancer. Recombinant TNFα is used as an immunostimulant under the INN tasonermin. TNFα can be produced ectopically in the setting of malignancy and parallels parathyroid hormone both in causing secondary hypercalcemia and in the cancers with which excessive production is associated.

This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 20.2 kDa. The protein migrates as 20-21 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

20.2 kDa
MABSol™ Biotinylated Mouse TNF-alpha, His,Avitag (active trimer) (MALS verified) (TNA-M82E9) is expressed from human HEK293 cells. It contains AA Leu 80 - Leu 235 (Accession # P06804).
-20°C
TNF-alpha
Mouse
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins
25 μg
Leu 80 - Leu 235
DIF,TNF-alpha,TNFA,TNFSF2,cachexin,cachectin,TNFα
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote